Assay Details
| Gene Symbol: RHEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285656.1 | NP_001272585.1 | 3 - 4 | 192 | 68 | |
| GenBank mRNA | HM569224.1 | - | 3 - 4 | 192 | 68 | |
More Information
Set Membership: |
|

RHEB
Genomic Map