Assay Details
| Gene Symbol: LDH-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285718.1 | NP_001272647.1 | 1 | 714 | 73 | |
| XM_013975786.2 | XP_013831240.2 | 6 - 7 | 969 | 73 | ||
| XM_018042654.1 | XP_017898143.1 | 6 - 7 | 946 | 73 | ||
| GenBank mRNA | GU144018.1 | - | 1 | 714 | 73 | |
| JO580468.1 | - | 6 - 7 | 906 | 73 | ||
More Information
Set Membership: |
|

LDH-A
Genomic Map