Assay Details
| Gene Symbol: KRT25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285766.1 | NP_001272695.1 | 2 - 3 | 568 | 72 | |
| GenBank mRNA | AY510111.1 | - | 2 - 3 | 568 | 72 | |
| KA352949.1 | - | 6 - 7 | 1276 | 72 | ||
More Information
Set Membership: |
|

KRT25
Genomic Map