Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289949.1 | NP_001276878.1 | 6 - 7 | 640 | 109 | |
| XM_018049973.1 | XP_017905462.1 | 6 - 7 | 752 | 109 | ||
| XM_018049974.1 | XP_017905463.1 | 6 - 7 | 770 | 109 | ||
| XM_018049975.1 | XP_017905464.1 | 6 - 7 | 751 | 109 | ||
| GenBank mRNA | KC537058.1 | - | 6 - 7 | 640 | 109 | |
More Information
Set Membership: |
|

MYOZ3
Genomic Map