Assay Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314307.1 | NP_001301236.1 | 3 - 4 | 495 | 60 | |
| XM_018047552.1 | XP_017903041.1 | 4 - 5 | 692 | 60 | ||
| GenBank mRNA | JX573540.1 | - | 3 - 4 | 495 | 60 | |
More Information
Set Membership: |
|

ADIPOR2
Genomic Map