Assay Details
| Gene Symbol: TLR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285574.1 | NP_001272503.1 | 1 - 2 | 195 | 108 | |
| GenBank mRNA | JF825527.1 | - | 1 - 2 | 195 | 108 | |
| JO519293.2 | - | 1 - 2 | 350 | 108 | ||
More Information
Set Membership: |
|

TLR4
Genomic Map