Assay Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285731.1 | NP_001272660.1 | 1 - 2 | 73 | 105 | |
| XM_018058851.1 | XP_017914340.1 | 1 - 2 | 111 | 105 | ||
| GenBank mRNA | GU338978.1 | - | 1 - 2 | 76 | 105 | |
| KA307331.1 | - | 3 - 4 | 1030 | 105 | ||
More Information
Set Membership: |
|

SDHD
Genomic Map