Assay Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285731.1 | NP_001272660.1 | 2 - 3 | 196 | 85 | |
| GenBank mRNA | GU338978.1 | - | 2 - 3 | 199 | 85 | |
| KA307331.1 | - | 2 - 3 | 907 | 85 | ||
More Information
Set Membership: |
|

SDHD
Genomic Map