Assay Details
| Gene Symbol: ADIPOQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287573.1 | NP_001274502.1 | 1 - 2 | 208 | 60 | |
| XM_005675149.3 | XP_005675206.1 | 2 - 3 | 271 | 60 | ||
| XM_018056531.1 | XP_017912020.1 | 3 - 4 | 314 | 60 | ||
| GenBank mRNA | JX573539.1 | - | 1 - 2 | 216 | 60 | |
| KF452236.1 | - | 1 - 2 | 208 | 60 | ||
More Information
Set Membership: |
|

ADIPOQ
Genomic Map