Assay Details
| Gene Symbol: HSPA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314233.1 | NP_001301162.1 | 5 - 6 | 1317 | 68 | |
| XM_005689565.3 | XP_005689622.1 | 6 - 7 | 1450 | 68 | ||
| GenBank mRNA | JO579495.1 | - | 3 - 4 | 930 | 68 | |
| JQ990218.1 | - | 5 - 6 | 1317 | 68 | ||
| JQ990219.1 | - | 5 - 6 | 1317 | 68 | ||
More Information
Set Membership: |
|

HSPA8
Genomic Map