Assay Details
| Gene Symbol: PROLACTIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285547.1 | NP_001272476.1 | 371 | 65 | ||
| GenBank mRNA | JF938531.1 | - | 374 | 65 | ||
| X76048.1 | - | 373 | 65 | |||
| X76049.1 | - | 373 | 65 | |||
More Information
Set Membership: |
|

PROLACTIN
Genomic Map