Assay Details
| Gene Symbol: HMOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285567.1 | NP_001272496.1 | 1 - 2 | 24 | 50 | |
| GenBank mRNA | JF731041.1 | - | 1 - 2 | 24 | 50 | |
| JO580392.1 | - | 1 - 2 | 194 | 50 | ||
| KA334415.1 | - | 1 - 2 | 176 | 50 | ||
More Information
Set Membership: |
|

HMOX1
Genomic Map