Assay Details
| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285587.1 | NP_001272516.1 | 2 - 3 | 113 | 95 | |
| GenBank mRNA | JT653927.1 | - | 2 - 3 | 153 | 95 | |
| X60763.1 | - | 2 - 3 | 113 | 95 | ||
More Information
Set Membership: |
|

CSN3
Genomic Map