Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285592.1 | NP_001272521.1 | 1 - 2 | 214 | 64 | |
| XM_018042146.1 | XP_017897635.1 | 3 - 4 | 620 | 64 | ||
| XM_018042147.1 | XP_017897636.1 | 3 - 4 | 498 | 64 | ||
| GenBank mRNA | JN561166.1 | - | 1 - 2 | 214 | 64 | |
| JO583826.1 | - | 2 - 3 | 420 | 64 | ||
More Information
Set Membership: |
|

BMPR1A
Genomic Map