Assay Details
| Gene Symbol: GABARAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285597.1 | NP_001272526.1 | 2 - 3 | 93 | 70 | |
| GenBank mRNA | HQ846829.1 | - | 2 - 3 | 93 | 70 | |
| JO633454.2 | - | 249 | 70 | |||
More Information
Set Membership: |
|

GABARAPL2
Genomic Map