Assay Details
| Gene Symbol: PRLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285669.1 | NP_001272598.1 | 5 - 6 | 683 | 73 | |
| XM_013972803.2 | XP_013828257.2 | 7 - 8 | 1542 | 73 | ||
| XM_013972804.2 | XP_013828258.2 | 7 - 8 | 927 | 73 | ||
| XM_013972806.2 | XP_013828260.2 | 6 - 7 | 1506 | 73 | ||
| XM_013972808.2 | XP_013828262.1 | 7 - 8 | 1550 | 73 | ||
| XM_018065466.1 | XP_017920955.1 | 7 - 8 | 1567 | 73 | ||
| XM_018065467.1 | XP_017920956.1 | 7 - 8 | 1575 | 73 | ||
| GenBank mRNA | GU075814.1 | - | 5 - 6 | 635 | 73 | |
| GU075815.1 | - | 5 - 6 | 635 | 73 | ||
| HQ667935.1 | - | 5 - 6 | 635 | 73 | ||
| JF966783.1 | - | 5 - 6 | 683 | 73 | ||
| JO499036.2 | - | 3 - 4 | 378 | 73 | ||
| JX087440.1 | - | 5 - 6 | 699 | 73 | ||
| KC906265.1 | - | 5 - 6 | 738 | 73 | ||
| KF850473.1 | - | 5 - 6 | 693 | 73 | ||
| KJ792812.1 | - | 5 - 6 | 683 | 73 | ||
| KJ792813.1 | - | 5 - 6 | 683 | 73 | ||
More Information
Set Membership: |
|

PRLR
Genomic Map