Assay Details
| Gene Symbol: CXCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285721.1 | NP_001272650.1 | 3 - 4 | 338 | 101 | |
| GenBank mRNA | AB099892.1 | - | 3 - 4 | 338 | 101 | |
| JO581549.1 | - | 3 - 4 | 457 | 101 | ||
More Information
Set Membership: |
|

CXCL10
Genomic Map