Assay Details
| Gene Symbol: B4GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285775.1 | NP_001272704.1 | 2 - 3 | 677 | 64 | |
| GenBank mRNA | HQ700335.1 | - | 2 - 3 | 677 | 64 | |
| JO496616.2 | - | 4 - 5 | 2011 | 64 | ||
More Information
Set Membership: |
|

B4GALT1
Genomic Map