Assay Details
| Gene Symbol: KANSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287026.1 | NP_001273955.1 | 6 - 7 | 931 | 59 | |
| NM_001287027.1 | NP_001273956.1 | 6 - 7 | 931 | 59 | ||
| XM_013963665.2 | XP_013819119.1 | 6 - 7 | 1556 | 59 | ||
| XM_013963666.2 | XP_013819120.1 | 6 - 7 | 1557 | 59 | ||
| GenBank mRNA | AY214590.1 | - | 5 - 6 | 1130 | 59 | |
| AY214591.1 | - | 5 - 6 | 1130 | 59 | ||
| AY214592.1 | - | 4 - 5 | 1015 | 59 | ||
| AY214593.1 | - | 4 - 5 | 1015 | 59 | ||
| AY245697.1 | - | 5 - 6 | 1130 | 59 | ||
| AY245698.1 | - | 5 - 6 | 1130 | 59 | ||
| JO404642.2 | - | 4 - 5 | 1024 | 59 | ||
| KA348946.1 | - | 4 - 5 | 1131 | 59 | ||
More Information
Set Membership: |
|

KANSL2
Genomic Map