Assay Details
| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287568.1 | NP_001274497.1 | 4 - 5 | 588 | 81 | |
| XM_013964451.2 | XP_013819905.1 | 5 - 6 | 649 | 81 | ||
| GenBank mRNA | JX573191.1 | - | 4 - 5 | 588 | 81 | |
More Information
Set Membership: |
|

MYOZ2
Genomic Map