Assay Details
| Gene Symbol: FABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287575.1 | NP_001274504.1 | 2 - 3 | 242 | 95 | |
| GenBank mRNA | JQ031285.1 | - | 2 - 3 | 242 | 95 | |
More Information
Set Membership: |
|

FABP2
Genomic Map