Assay Details
| Gene Symbol: FABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287575.1 | NP_001274504.1 | 3 - 4 | 343 | 91 | |
| GenBank mRNA | JQ031285.1 | - | 3 - 4 | 343 | 91 | |
More Information
Set Membership: |
|

FABP2
Genomic Map