Assay Details
| Gene Symbol: LOC102189356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314188.1 | NP_001301117.1 | 2 - 3 | 326 | 57 | |
| XM_005696526.3 | XP_005696583.1 | 2 - 3 | 439 | 57 | ||
| GenBank mRNA | AB008754.1 | - | 2 - 3 | 326 | 57 | |
| JO530072.2 | - | 2 - 3 | 460 | 57 | ||
| JO579669.1 | - | 1 - 2 | 145 | 57 | ||
More Information
Set Membership: |
|

LOC102189356
Genomic Map