Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285539.1 | NP_001272468.1 | 6 - 7 | 615 | 84 | |
| GenBank mRNA | JO517063.2 | - | 1 - 2 | 76 | 84 | |
| X58471.1 | - | 6 - 7 | 587 | 84 | ||
| Z19569.1 | - | 6 - 7 | 615 | 84 | ||
More Information
Set Membership: |
|

PAEP
Genomic Map