Assay Details
| Gene Symbol: IL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285681.1 | NP_001272610.1 | 2 - 3 | 245 | 64 | |
| GenBank mRNA | AY648947.1 | - | 2 - 3 | 203 | 64 | |
| U34273.1 | - | 2 - 3 | 247 | 64 | ||
More Information
Set Membership: |
|

IL4
Genomic Map