Assay Details
| Gene Symbol: GNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285690.1 | NP_001272619.1 | 1 - 2 | 228 | 63 | |
| GenBank mRNA | JO373133.2 | - | 1 - 2 | 240 | 63 | |
| U17694.1 | - | 1 - 2 | 228 | 63 | ||
More Information
Set Membership: |
|

GNS
Genomic Map