Assay Details
| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285714.1 | NP_001272643.1 | 3 - 4 | 711 | 107 | |
| GenBank mRNA | GU084153.1 | - | 3 - 4 | 711 | 107 | |
More Information
Set Membership: |
|

KRT26
Genomic Map