Assay Details
Gene Symbol: CASP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286089.1 | NP_001273018.1 | 5 - 6 | 615 | 73 | |
XM_018041755.1 | XP_017897244.1 | 6 - 7 | 721 | 73 | ||
GenBank mRNA | JN986829.1 | - | 5 - 6 | 615 | 73 | |
JO416587.2 | - | 6 - 7 | 678 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
