Assay Details
| Gene Symbol: FASLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291820.1 | NP_001278749.1 | 1 - 2 | 352 | 74 | |
| GenBank mRNA | KJ136646.1 | - | 1 - 2 | 352 | 74 | |
More Information
Set Membership: |
|

FASLG
Genomic Map