Assay Details
| Gene Symbol: ENO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314158.1 | NP_001301087.1 | 10 - 11 | 1235 | 68 | |
| XM_018047555.1 | XP_017903044.1 | 11 - 12 | 68 | |||
| GenBank mRNA | JN887466.1 | - | 10 - 11 | 1235 | 68 | |
| JO633950.2 | - | 11 - 12 | 68 | |||
More Information
Set Membership: |
|

ENO2
Genomic Map