Assay Details
| Gene Symbol: OPRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314237.1 | NP_001301166.1 | 3 - 4 | 722 | 92 | |
| XM_018058073.1 | XP_017913562.1 | 3 - 4 | 783 | 92 | ||
| GenBank mRNA | AB900618.1 | - | 3 - 4 | 722 | 92 | |
More Information
Set Membership: |
|

OPRK1
Genomic Map