Assay Details
| Gene Symbol: ODF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314333.3 | NP_001301262.3 | 1 - 2 | 420 | 72 | |
| GenBank mRNA | JX067553.2 | - | 1 - 2 | 424 | 72 | |
More Information
Set Membership: |
|

ODF1
Genomic Map