Assay Details
| Gene Symbol: GFI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285593.1 | NP_001272522.1 | 4 - 5 | 653 | 49 | |
| GenBank mRNA | JN561130.1 | - | 4 - 5 | 653 | 49 | |
| JN662390.1 | - | 4 - 5 | 653 | 49 | ||
| JR122562.1 | - | 400 | 49 | |||
| JX627316.1 | - | 4 - 5 | 653 | 49 | ||
More Information
Set Membership: |
|

GFI1B
Genomic Map