Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285619.1 | NP_001272548.1 | 3 - 4 | 436 | 66 | |
| GenBank mRNA | AF325499.1 | - | 3 - 4 | 632 | 66 | |
| AF339909.1 | - | 3 - 4 | 608 | 66 | ||
| GU947654.1 | - | 3 - 4 | 436 | 66 | ||
| JO634430.2 | - | 4 - 5 | 3946 | 66 | ||
| JX854036.1 | - | 3 - 4 | 436 | 66 | ||
| KA308300.1 | - | 3 - 4 | 871 | 66 | ||
| KT315581.1 | - | 3 - 4 | 537 | 66 | ||
More Information
Set Membership: |
|

SCD
Genomic Map