Assay Details
| Gene Symbol: PRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285622.1 | NP_001272551.1 | 4 - 5 | 575 | 88 | |
| XM_018061647.1 | XP_017917136.1 | 4 - 5 | 682 | 88 | ||
| GenBank mRNA | AB231295.1 | - | 4 - 5 | 575 | 88 | |
More Information
Set Membership: |
|

PRP1
Genomic Map