Assay Details
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285759.1 | NP_001272688.1 | 2 - 3 | 259 | 61 | |
| GenBank mRNA | AY389343.1 | - | 2 - 3 | 259 | 61 | |
| GQ369727.1 | - | 2 - 3 | 210 | 61 | ||
| JN377801.1 | - | 1 - 2 | 165 | 61 | ||
More Information
Set Membership: |
|

CYM
Genomic Map