Assay Details
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285759.1 | NP_001272688.1 | 4 - 5 | 500 | 75 | |
| GenBank mRNA | AY389343.1 | - | 4 - 5 | 500 | 75 | |
| EF199763.1 | - | 4 - 5 | 463 | 75 | ||
| GQ369727.1 | - | 4 - 5 | 451 | 75 | ||
More Information
Set Membership: |
|

CYM
Genomic Map