Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285764.1 | NP_001272693.1 | 5 - 6 | 945 | 65 | |
| XM_005694718.3 | XP_005694775.3 | 5 - 6 | 1384 | 65 | ||
| GenBank mRNA | HM138664.1 | - | 5 - 6 | 1106 | 65 | |
| HM138666.1 | - | 5 - 6 | 1106 | 65 | ||
| HM852511.1 | - | 5 - 6 | 945 | 65 | ||
| JQ976292.1 | - | 5 - 6 | 945 | 65 | ||
More Information
Set Membership: |
|

FST
Genomic Map