Assay Details
| Gene Symbol: CTSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314163.1 | NP_001301092.1 | 6 - 7 | 497 | 59 | |
| GenBank mRNA | JO579420.1 | - | 6 - 7 | 617 | 59 | |
| KJ650499.1 | - | 6 - 7 | 497 | 59 | ||
More Information
Set Membership: |
|

CTSH
Genomic Map