Assay Details
| Gene Symbol: PRNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314247.1 | NP_001301176.1 | 1 | 526 | 78 | |
| XM_005688157.3 | XP_005688214.2 | 3 | 750 | 78 | ||
| GenBank mRNA | JO408879.2 | - | 3 | 1060 | 78 | |
| KP715243.1 | - | 1 | 526 | 78 | ||
| KP715244.1 | - | 1 | 526 | 78 | ||
| KP715245.1 | - | 1 | 526 | 78 | ||
More Information
Set Membership: |
|

PRNP
Genomic Map