Assay Details
| Gene Symbol: LIN28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314177.1 | NP_001301106.1 | 1 | 414 | 103 | |
| GenBank mRNA | KJ755856.1 | - | 1 | 414 | 103 | |
| Gene Symbol: LIN28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067111.1 | XP_017922600.1 | 3 - 4 | 414 | 103 | |
More Information
Set Membership: |
|

LIN28A
Genomic Map