Assay Details
| Gene Symbol: IL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058383.1 | XP_017913872.1 | 4 - 5 | 643 | 59 | |
| GenBank mRNA | JT683122.1 | - | 4 - 5 | 839 | 59 | |
More Information
Set Membership: |
|

IL7
Genomic Map