Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694738.3 | XP_005694795.1 | 2 - 3 | 273 | 89 | |
| GenBank mRNA | JO443631.2 | - | 2 - 3 | 346 | 89 | |
More Information
Set Membership: |
|

CCL28
Genomic Map