Assay Details
| Gene Symbol: ARFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059469.1 | XP_017914958.1 | 5 - 6 | 715 | 67 | |
| XM_018059470.1 | XP_017914959.1 | 2 - 3 | 287 | 67 | ||
| GenBank mRNA | JO599511.2 | - | 3 - 4 | 1068 | 67 | |
More Information
Set Membership: |
|

ARFIP2
Genomic Map