Assay Details
| Gene Symbol: CLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056496.1 | XP_017911985.1 | 3 - 4 | 586 | 80 | |
| GenBank mRNA | JO427129.1 | - | 2 - 3 | 2345 | 80 | |
More Information
Set Membership: |
|

CLN5
Genomic Map