Assay Details
| Gene Symbol: CCND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680985.3 | XP_005681042.1 | 4 - 5 | 1042 | 63 | |
| GenBank mRNA | JO375303.2 | - | 272 | 63 | ||
More Information
Set Membership: |
|

CCND2
Genomic Map