Assay Details
| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057383.1 | XP_017912872.1 | 4 - 5 | 1434 | 54 | |
| XM_018057385.1 | XP_017912874.1 | 4 - 5 | 1223 | 54 | ||
| XM_018057386.1 | XP_017912875.1 | 5 - 6 | 942 | 54 | ||
| XM_018057387.1 | XP_017912876.1 | 4 - 5 | 665 | 54 | ||
| XM_018057388.1 | XP_017912877.1 | 2 - 3 | 394 | 54 | ||
| XM_018057389.1 | XP_017912878.1 | 2 - 3 | 559 | 54 | ||
| GenBank mRNA | JO604740.2 | - | 3 - 4 | 740 | 54 | |
| JO604743.2 | - | 3 - 4 | 851 | 54 | ||
More Information
Set Membership: |
|

PANK2
Genomic Map