Assay Details
Gene Symbol: ARL5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693735.3 | XP_005693792.1 | 1 - 2 | 115 | 61 | |
GenBank mRNA | JO340142.2 | - | 5 - 6 | 1047 | 61 | |
JO598516.2 | - | 1 - 2 | 146 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
