Assay Details
| Gene Symbol: LCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692166.1 | XP_005692223.1 | 5 - 6 | 1017 | 72 | |
| XM_018062176.1 | XP_017917665.1 | 5 - 6 | 928 | 72 | ||
| GenBank mRNA | KA345110.1 | - | 4 - 5 | 954 | 72 | |
More Information
Set Membership: |
|

LCAT
Genomic Map