Assay Details
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683986.2 | XP_005684043.1 | 1 - 2 | 167 | 104 | |
| XM_018052283.1 | XP_017907772.1 | 1 - 2 | 131 | 104 | ||
| XM_018052284.1 | XP_017907773.1 | 1 - 2 | 130 | 104 | ||
More Information
Set Membership: |
|

SFTPC
Genomic Map